Mangandinatrium-EDTA-Hydrat
Lieferant: Thermo Scientific
Synonyme:
EDTA disodium manganese, Manganese Disodium Ethylenediaminetetraacetate, Manganese(II) disodium 2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetate
40519.06
40519.14
40519.06EA
39.4
EUR
40519.06
40519.14
Mangandinatrium-EDTA-Hydrat
Ethylenediaminetetraacetic acid manganese disodium salt hydrate
CAS No.: 15375-84-5
Molecular Formula: (NaO2CCH2)2NCH2CH2N(CH2CO2)2Mn.xH2O
Formula Weight: 389.13(anhy)
Physical Form: Powder
Odour: Odourless
MDL No.: MFCD00661156
Applications: Metal chelating compound
Formel:
C₁₀H₁₆N₂O₈ Molecular Weight: 389,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054451 CAS-Nummer: 15375-84-5 EINECS: 239-407-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...