Samarium(III) 2,4-pentanedionate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Scientific
40665.06
40665.14
40665.06EA
37.3
EUR
40665.06
40665.14
Samarium(III) 2,4-pentanedionate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Samarium(III) 2,4-pentanedionate hydrate
Purity: 99.9%
CAS No.: 64438-53-5
Molecular Formula: C15H21O6Sm.xH2O
Formula Weight: 447.68(anhy)
Physical Form: Powder
MDL No.: MFCD00792904
Formel:
C₁₅H₂₁O₆Sm·xH₂O Molecular Weight: 447,68 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00792904 CAS-Nummer: 64438-53-5 EINECS: 238-632-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...