Zinc tert-butoxide
Lieferant: Thermo Scientific
|
Achtung
|
40718.03
40718.MD
40718.03EA
234
EUR
40718.03
40718.MD
Zinc tert-butoxide
Zinc tert-butoxide
CAS No.: 4278-43-7
Molecular Formula: C8H18O2Zn
Formula Weight: 211.61
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White to pale yellow
Odour: Odourless
UN No.: UN3181
MDL No.: MFCD00145545
Formel:
C₈H₁₈O₂Zn Lagertemperatur: Raumtemperatur |
MDL:
MFCD00145545 CAS-Nummer: 4278-43-7 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...