Germanium-n-butoxide ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
40816.03
40816.06
40816.14
40816.03EA
48.3
EUR
40816.03
40816.06
40816.14
Germanium-n-butoxide ≥97%
Germanium-n-butoxide
Purity: 97+%
CAS No.: 25063-27-8
Molecular Formula: C16H36GeO4
Formula Weight: 365.05
Flash Point: 112°(234°F)
Physical Form: Liquid
Density: 1.017
Refractive Index: 1.4255
Sensitivity: Moisture Sensitive
Boiling Point: 143°/8mm
MDL No.: MFCD00015347
Applications: Oxide films
Formel:
C₁₆H₃₆GeO₄ Molecular Weight: 365,05 g/mol Siedepunkt: 143 °C (8 mmHg) Dichte: 1,017 Flammpunkt: 112 °C (234 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015347 CAS-Nummer: 25063-27-8 EINECS: 246-595-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...