Carbonylhydridotris(triphenylphosphine)iridium(I) (≥18,6% Ir)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Hydridocarbonyltris(triphenylphosphine)iridium(I)

41005.03 41005.06
41005.03EA 0 EUR
41005.03 41005.06
Carbonylhydridotris(triphenylphosphine)iridium(I) (≥18,6% Ir)
Carbonylhydridotris(triphenylphosphine)iridium(I)

CAS No.: 17250-25-8 Molecular Formula: C55H46IrOP3 Formula Weight: 1008.11 Physical Form: Powder Sensitivity: Moisture Sensitive Appearance: Pale yellow Melting Point: 170° Solubility: Soluble in chloroform and toluene MDL No.: MFCD00015525

Formel: C₅₅H₄₆IrOP₃
Molecular Weight: 1008.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00015525
CAS-Nummer: 17250-25-8
EINECS: 241-282-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR