Tellurium(IV) ethoxide ≥85% remainder ethanol, tech.

Lieferant: Thermo Scientific
Achtung

41105.04 41105.09
41105.04EA 134 EUR
41105.04 41105.09
Tellurium(IV) ethoxide ≥85% remainder ethanol, tech.
Tellurium(IV) ethoxide

Purity: 85% CAS No.: 2017-01-8 Molecular Formula: Te(OC2H5)4 Formula Weight: 307.84 Flash Point: 27°(81°F) Physical Form: Liquid Density: 1.52 Sensitivity: Moisture and Light Sensitive Odour: Alcohol-like Melting Point: 100° dec. Boiling Point: 88-90°/2mm Solubility: Decomposes in water to TeO2 UN No.: UN1993 MDL No.: MFCD00270937

Formel: Te(OC₂H₅)₄
Molecular Weight: 307,84 g/mol
Siedepunkt: 88…90 °C (2 mmHg)
Schmelzpunkt: >100 °C (Zersetzung)
Dichte: 1,52
Flammpunkt: 27 °C (81 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00270937
CAS-Nummer: 2017-01-8
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR