Fullerene, hydroxylated, C60(OH)n, Pulver
Lieferant: Thermo Scientific
|
Achtung
|
41182.MB
41182.MC
41182.MBEA
217
EUR
41182.MB
41182.MC
Fullerene, hydroxylated, C60(OH)n, Pulver
Fullerene, hydroxylated, C60(OH)n
UN No.: UN1325
MDL No.: MFCD06202029
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD06202029 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...