Cobalt(II)-phthalocyanin
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Cobalt(II) phthalocyanine, Phthalocyanine Cobalt(II), 29H,31H-Phthalocyanine, cobalt complex
41496.03
41496.06
41496.14
41496.03EA
36.4
EUR
41496.03
41496.06
41496.14
Cobalt(II)-phthalocyanin
Cobalt(II) Phthalocyanine
CAS No.: 3317-67-7
Molecular Formula: C32H16N8Co
Formula Weight: 571.47
Physical Form: Powder
Appearance: Dark blue to purple
Odour: Odourless
MDL No.: MFCD00010718
Formel:
C₃₂H₁₆CoN₈ Molecular Weight: 571,47 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010718 CAS-Nummer: 3317-67-7 EINECS: 222-012-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...