Naphthalene-1,3,6-trisulphonic acid trisodium salt hydrate

Lieferant: Thermo Scientific
Achtung

Synonyme: 1,3,6-Naphthalenetrisulphonic acid, trisodium salt hydrate

41715.22 41715.36
41715.22EA 41.6 EUR
41715.22 41715.36
Naphthalene-1,3,6-trisulphonic acid trisodium salt hydrate
Naphthalene-1,3,6-trisulphonic acid trisodium salt hydrate

CAS No.: 123409-01-8 Molecular Formula: C10H5Na3O9S3.xH2O Formula Weight: 434.31(anhy) Physical Form: Powder MDL No.: MFCD00149263

Formel: C₁₀H₅Na₃O₉S₃·xH₂O
Molecular Weight: 434,31 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149263
CAS-Nummer: 123409-01-8
EINECS: 225-962-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR