Hexane-1,6-bis(tri-n-butylammonium) dihydroxide 20% (w/w) in wässriger Lösung
Lieferant: Thermo Scientific
Gefahr
Synonyme: Bisquat dihydroxide, N,N,N,N',N',N'-Hexa-n-butylhexamethylenediammonium dihydroxide
41725.AD 41725.AK 41725.K2
41725.ADEA 68.7 EUR
41725.AD 41725.AK 41725.K2
Hexane-1,6-bis(tri-n-butylammonium) dihydroxide 20% (w/w) in wässriger Lösung
Hexane-1,6-bis(tri-n-butylammonium) dihydroxide
CAS No.: 69762-88-5 Molecular Formula: C30H68N2O2 Formula Weight: 488.87 Physical Form: Liquid UN No.: UN3267 MDL No.: MFCD00216633
Formel: C₃₀H₆₈N₂O₂
Molecular Weight: 488,87 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00216633
CAS-Nummer: 69762-88-5
UN: 3267
ADR: 8,III