Bismuth tungsten oxide ≥99,9% (Metall-Basis)
Lieferant: Thermo Scientific
41854.14
41854.22
41854.14EA
141
EUR
41854.14
41854.22
Bismuth tungsten oxide ≥99,9% (Metall-Basis)
Bismuth tungsten oxide
Purity: 99.9%
CAS No.: 13595-87-4
Molecular Formula: Bi2(WO4)3
Formula Weight: 1161.48
Physical Form: -200 Mesh Powder
Odour: Odourless
MDL No.: MFCD00046190
Formel:
Bi₂(WO₄)₃ Molecular Weight: 1161,48 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00046190 CAS-Nummer: 13595-87-4 EINECS: 237-033-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...