Polyimide, Film, Dicke 0.013 mm

Lieferant: Thermo Scientific

41897.GH 41897.GT 41897.HB
41897.GHEA 200 EUR
41897.GH 41897.GT 41897.HB
Polyimide, Film, Dicke 0.013 mm
Polyimide

CAS No.: 62929-02-6 Physical Form: ≈0.1846g/100x100mm MDL No.: MFCD00677744

Formel: C₃₅H₂₈N₂O₇
Lagertemperatur: Raumtemperatur
MDL: MFCD01868102
CAS-Nummer: 62929-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR