Polyimide, Film, Breite 20 mm, Dicke 0.025 mm

Lieferant: Thermo Scientific

41898.3Z
41898.3ZEA 0 EUR
41898.3Z
Polyimide, Film, Breite 20 mm, Dicke 0.025 mm
Polyimide

CAS No.: 62929-02-6 MDL No.: MFCD00677744

Formel: C₃₅H₂₈N₂O₇
Lagertemperatur: Raumtemperatur
MDL: MFCD01868102
CAS-Nummer: 62929-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR