Magnesium acrylate

Lieferant: Thermo Scientific
Achtung

42002.06 42002.14 42002.22
42002.06EA 26.8 EUR
42002.06 42002.14 42002.22
Magnesium acrylate
Magnesium acrylate

CAS No.: 5698-98-6 Molecular Formula: C6H6MgO4 Formula Weight: 166.42 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Melting Point: 220° dec. MDL No.: MFCD00068285

Formel: C₆H₆MgO₄
Schmelzpunkt: >220 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00068285
CAS-Nummer: 5698-98-6
EINECS: 227-177-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR