Indium oxide, tin doped (∼20% Oxide) (w/w) polimerische Anzeichen
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
42229.09
42229.14
42229.09EA
574
EUR
42229.09
42229.14
Indium oxide, tin doped (∼20% Oxide) (w/w) polimerische Anzeichen
Indium oxide, tin doped
CAS No.: 1312-43-2
Physical Form: Viscous liquid
Solubility: Soluble in hexane and toluene. Insoluble in alcohols
UN No.: UN1993
MDL No.: MFCD00011060
Applications: Preparation of transparent electrical conducting electrodes
Flammpunkt:
13 °C (55 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011060 UN: 1993 ADR: 3 III |