Amberlite® IRA-67 Ionenaustauscher

Lieferant: Thermo Fisher Scientific

42253.30 42253.A1 42253.A7
42253.30EA 49.3 EUR
42253.30 42253.A1 42253.A7
Amberlite® IRA-67 Ionenaustauscher
Amberlite® IRA-67

CAS No.: 80747-90-6 Odour: Amine-like MDL No.: MFCD00145567

Lagertemperatur: Raumtemperatur
MDL: MFCD00145567
CAS-Nummer: 80747-90-6
Merck Index: 14,382

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR