Apatite natürlich vorkommendes Mineral, Körner ∼ 0.06-0.19 in

Lieferant: Thermo Scientific
Achtung

Synonyme: Fluorapatite, Apatite, Calcium fluorophosphate

42535.03 42535.06 42535.14
42535.03EA 30.1 EUR
42535.03 42535.06 42535.14
Apatite natürlich vorkommendes Mineral, Körner ∼ 0.06-0.19 in
Apatite

Apatite is one of a few minerals produced and used by biological micro-environmental systems. The primary use of apatite is in the manufacture of fertilizer, it is a source of phosphorus. It is occasionally used as a gemstone. Green and blue varieties in finely divided form, are pigments with excellent covering power.


Sp. Gr. 3.1-3.2. Store away from strong oxidizing agents. Store in cool, dry conditions.


Applications: Grain mounts, XRD and microprobe standards, identification of unknown minerals

Formel: Ca₅FO₁₂P₃
Molecular Weight: 504,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00285511
CAS-Nummer: 1306-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR