Pyrite (FeS₂) natürlich vorkommendes Mineral, Körner ∼1.5-4.8mm (0.06-0.19in)

Lieferant: Thermo Fisher Scientific

42633.03 42633.06 42633.14
42633.03EA 36.6 EUR
42633.03 42633.06 42633.14
Pyrite (FeS₂) natürlich vorkommendes Mineral, Körner ∼1.5-4.8mm (0.06-0.19in)
Pyrite (FeS₂)

For grain mounts, XRD and microprobe standards, identification of unknown minerals
Hardness: 6-6.5


CAS No.: 1309-36-0 Molecular Formula: FeS2 Formula Weight: 119.98 Density: 5.02 Refractive Index: 3.08 MDL No.: MFCD00064690


Applications: For grain mounts, XRD and microprobe standards, identification of unknown minerals

Formel: FeS₂
Molecular Weight: 119.965 g/mol
Dichte: 5.02
Lagertemperatur: Raumtemperatur
MDL: MFCD00064690
CAS-Nummer: 1309-36-0
EINECS: 215-167-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR