Calcium-3-hydroxy-3-methylbutyrate hydrate ≥97% (durch titrimetrische Analyse), weißes Pulver
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Hydroxy-3-methylbutyric Acid Calcium Salt
42721.18
42721.30
42721.A1
42721.18EA
87.8
EUR
42721.18
42721.30
42721.A1
Calcium-3-hydroxy-3-methylbutyrate hydrate ≥97% (durch titrimetrische Analyse), weißes Pulver
Calcium-3-hydroxy-3-methylbutyrate hydrate
Purity: 97+%
CAS No.: 135236-72-5
Molecular Formula: C10H18CaO6.xH2O
Formula Weight: 274.33(anhy)
Physical Form: Powder
Appearance: White
MDL No.: MFCD01318562
Formel:
C₁₀H₁₈CaO₆·H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD01321171 CAS-Nummer: 135236-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...