Tetraammineplatinum(II) hydroxide-Lösung (8 - 11% Pt) (w/w)
Lieferant: Thermo Scientific
Achtung
42918.NB 42918.NH
42918.NBEA 1186 EUR
42918.NB 42918.NH
Tetraammineplatinum(II) hydroxide-Lösung (8 - 11% Pt) (w/w)
Tetraammineplatinum(II) hydroxide
CAS No.: 38201-97-7 Molecular Formula: H14N4O2Pt Formula Weight: 297.23 Physical Form: Liquid Appearance: Colourless Odour: Odourless MDL No.: MFCD00050044
Formel: H₁₄N₄O₂Pt
Molecular Weight: 297,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00050044
CAS-Nummer: 38201-97-7
EINECS: 253-823-4