Lithium isopropoxide ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
unter Argon abgefüllt
42930.04
42930.09
42930.04EA
81.6
EUR
42930.04
42930.09
Lithium isopropoxide ≥99,9% (Metall-Basis)
Lithium isopropoxide
Purity: 99.9%
CAS No.: 2388-10-5
Molecular Formula: C3H7LiO
Formula Weight: 66.03
Physical Form: Powder, Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: White to pale yellow
Beilstein Reference: 3618614
UN No.: UN3206
MDL No.: MFCD00048286
Formel:
C₃H₇LiO Molecular Weight: 66.028 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00048286 CAS-Nummer: 2388-10-5 UN: 3206 ADR: 4.2 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...