Iron(III) isopropoxide 2,5% (w/v) in 2-Propanol (IPA)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Iron(III) i-propoxide
unter Argon abgefüllt
43024.AC
43024.AA
43024.ACEA
117
EUR
43024.AC
43024.AA
Iron(III) isopropoxide 2,5% (w/v) in 2-Propanol (IPA)
Iron(III) isopropoxide
CAS No.: 14995-22-3
Molecular Formula: Fe[OCH(CH3)2]3
Formula Weight: 233.11
Flash Point: 12°(54°F)
Physical Form: Liquid, Packaged under Argon
Density: 0.79
Sensitivity: Moisture Sensitive
Appearance: Red-brown
Odour: Alcohol-like
Boiling Point: 82°
UN No.: UN1219
MDL No.: MFCD00070436
Formel:
C₉H₂₁FeO₃ Molecular Weight: 233,109 g/mol Siedepunkt: 82 °C Dichte: 0,79 Flammpunkt: 12 °C (54 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070436 CAS-Nummer: 14995-22-3 UN: 1219 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...