Benzyl-acrylat 98% stabilisiert
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Acrylic Acid Benzyl Ester
Stabilisierung:
Stabilisiert mit 4-Methoxyphenol (MEHQ) 150 ppm
43204.30
43204.18
43204.09
43204.30EA
673
EUR
43204.30
43204.18
43204.09
Benzyl-acrylat 98% stabilisiert
Benzyl acrylate
Stab. with ca 150ppm 4-methoxyphenol
Purity: 97%
CAS No.: 2495-35-4
Molecular Formula: C10H10O2
Formula Weight: 162.19
Physical Form: Liquid
Density: 1.06
Refractive Index: 1.5180
Appearance: Colourless
Boiling Point: 111°/8mm
Solubility: Soluble in organic solvents
UN No.: UN3082
MDL No.: MFCD00048147
Applications: Preparation of high refractive index polyacrylates
Formel:
C₁₀H₁₀O₂ Molecular Weight: 162,19 g/mol Siedepunkt: 111 °C (8 mmHg) Dichte: 1,06 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00048147 CAS-Nummer: 2495-35-4 EINECS: 219-673-9 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...