DMS (Dimethyl suberimidate dihydrochloride)
Lieferant: Thermo Scientific
Achtung
Synonyme: DMS
43244.09 43244.04
43244.09EA 196 EUR
43244.09 43244.04
DMS (Dimethyl suberimidate dihydrochloride)
DMS (Dimethyl suberimidate dihydrochloride)
CAS No.: 34490-86-3 Molecular Formula: C10H20N2O2.2HCl Formula Weight: 273.20 Physical Form: Crystalline Sensitivity: Moisture Sensitive Melting Point: 213-214° Solubility: Soluble in water MDL No.: MFCD00012574
Applications: Crosslinking of proteins and poly(allylamine)
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 273,20 g/mol
Schmelzpunkt: 213…214 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012574
CAS-Nummer: 34490-86-3
EINECS: 252-060-4