Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), Pulver -20+35 mesh Type 316-L
Lieferant: Thermo Scientific
|
Gefahr
|
43460.A3
43460.36
43460.A3EA
331
EUR
43460.A3
43460.36
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), Pulver -20+35 mesh Type 316-L
Stainless steel
Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
MDL No.: MFCD00148499
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...