5-Methylcytosine ≥97%

Lieferant: Thermo Scientific

Synonyme: 4-Amino-5-methylpyrimidin-2(1H)-one

43492.MC 43492.ME
43492.MCEA 122 EUR
43492.MC 43492.ME
5-Methylcytosine ≥97%
5-Methylcytosine

Purity: 97% CAS No.: 554-01-8 Molecular Formula: C5H7N3O Formula Weight: 125.13 Physical Form: Crystalline MDL No.: MFCD00233537

Formel: C₅H₇N₃O
Molecular Weight: 125,131 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00233537
CAS-Nummer: 554-01-8
EINECS: 209-058-3
Merck Index: 14,6050

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR