Kristallviolett Lacton
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-(Dimethylamino)-3,3-bis[p-(dimethylamino)phenyl]phthalide, 3,3-Bis(p-dimethylaminophenyl)-6-dimethylaminophthalide
43710.14
43710.06
43710.14EA
260
EUR
43710.14
43710.06
Kristallviolett Lacton
Crystal violet lactone
CAS No.: 1552-42-7
Molecular Formula: C26H29N3O2
Formula Weight: 415.53
Physical Form: Powder
Melting Point: 180°-183°
MDL No.: MFCD00070611
Formel:
C₂₆H₂₉N₃O₂ Molecular Weight: 415,54 g/mol Schmelzpunkt: 180 °C…183 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070611 CAS-Nummer: 1552-42-7 EINECS: 216-293-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...