Polydimethylsiloxane, hydroxy terminated, M.W. 4200
Lieferant: Thermo Scientific
Synonyme: Polysiloxanes, di-Me, hydroxy-terminated
43769.K2 43769.K4 43769.AK 43769.AD
43769.K2EA 217 EUR
43769.K2 43769.K4 43769.AK 43769.AD
Polydimethylsiloxane, hydroxy terminated, M.W. 4200
Polydimethylsiloxane, hydroxy terminated
CAS No.: 70131-67-8 Molecular Formula: HO[-Si(CH3)2O-]nH Physical Form: Liquid Density: 0.97 MDL No.: MFCD01325012
Formel: [Si(CH₃)₂O]n
Molecular Weight: 45,092 g/mol
Dichte: 0,97
Lagertemperatur: Raumtemperatur
MDL: MFCD01325012
CAS-Nummer: 70131-67-8