Ethyl-N-Phenylformimidat ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-Phenylformimidic Acid Ethyl Ester, Ethyl N-phenylformimidate, N-Ethoxymethyleneaniline

43788.14 43788.06
43788.14EA 83.8 EUR
43788.14 43788.06
Ethyl-N-Phenylformimidat ≥97%
Ethyl-N-phenylformimidate

Purity: 97% CAS No.: 6780-49-0 Molecular Formula: C9H11NO Formula Weight: 149.19 Physical Form: Liquid Sensitivity: Air and Moisture Sensitive Appearance: Colourless liquid Boiling Point: 101-102°/15mm MDL No.: MFCD00043655

Formel: C₉H₁₁NO
Molecular Weight: 149,19 g/mol
Siedepunkt: 101…102 °C (15 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00043655
CAS-Nummer: 6780-49-0
EINECS: 229-841-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR