Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate

Lieferant: Thermo Scientific
Gefahr

Synonyme: Bis(norbornadiene)rhodium(I) tetrafluoroborate, Bis(norbornadiene)rhodium (I) tetrafluoroborate

44036.03 44036.06
44036.03EA 714 EUR
44036.03 44036.06
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate

CAS No.: 36620-11-8 Molecular Formula: [Rh(C7H8)2]BF4 Formula Weight: 373.99 Physical Form: Powder Sensitivity: Air and Moisture Sensitive MDL No.: MFCD00671775

Formel: C₁₄H₁₆BF₄Rh
Molecular Weight: 365,92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00671775
CAS-Nummer: 36620-11-8
UN: 1759
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR