Ammonium magnesium phosphate hydrate ≥99,999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

44053.18 44053.30
44053.18EA 165 EUR
44053.18 44053.30
Ammonium magnesium phosphate hydrate ≥99,999% (Metall-Basis)
Ammonium magnesium phosphate hydrate

Purity: 99.999% CAS No.: 7785-21-9 Molecular Formula: NH4MgPO4.xH2O Formula Weight: 137.32(anhy) Physical Form: Powder Density: 1.711 MDL No.: MFCD00050358

Formel: NH₄MgPO₄·xH₂O
Dichte: 1,711
Lagertemperatur: Raumtemperatur
MDL: MFCD00050358
CAS-Nummer: 7785-21-9
EINECS: 232-075-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR