Rhodium : Palladium on activated carbon (10:1), Pulver
Lieferant: Thermo Scientific
|
Achtung
|
44124.06
44124.14
44124.22
44124.06EA
0
EUR
44124.06
44124.14
44124.22
Rhodium : Palladium on activated carbon (10:1), Pulver
Rhodium : Palladium on activated carbon
Odour: Odourless
UN No.: UN1325
MDL No.: MFCD03792580
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD03792580 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...