N,N'-Bis(acryloyl)cystamine ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N,N'-(Disulphanediylbis(ethane-2,1-diyl))diacrylamide
unter Argon abgefüllt
44132.03
44132.06
44132.14
44132.03EA
99.9
EUR
44132.03
44132.06
44132.14
N,N'-Bis(acryloyl)cystamine ≥97%
N,N'-Bis(acryloyl)cystamine
Purity: 98%
CAS No.: 60984-57-8
Molecular Formula: C10H16N2O2S2
Formula Weight: 260.37
Storage Temperature: -30°C to -10°C
Physical Form: Powder, Packaged under argon
Appearance: White
Melting Point: 123-124°
MDL No.: MFCD00036225
Applications: Used as a reversible cross-linker for polyacrylamide gels
Formel:
C₁₀H₁₆N₂O₂S₂ Molecular Weight: 260,37 g/mol Schmelzpunkt: 123…124 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00036225 CAS-Nummer: 60984-57-8 EINECS: 262-546-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...