Poly(vinyl formal)

Lieferant: Thermo Scientific

Synonyme: Polyvinylformal, Formvar

44190.14 44190.22
44190.14EA 89.8 EUR
44190.14 44190.22
Poly(vinyl formal)
Poly(vinyl formal)

CAS No.: 9003-33-2 Physical Form: Granular/ powder Density: 1.23 Solubility: Soluble in acetone, chloroform, dioxane, THF MDL No.: MFCD00084463

Formel: (C₃H₆O₂)n
Dichte: 1,23
Lagertemperatur: Raumtemperatur
MDL: MFCD00084463
CAS-Nummer: 9003-33-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR