2,2'-Bicinchoninic acid disodium salt
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Sodium [2,2'-biquinoline]-4,4'-dicarboxylate, BCA disodium salt, Disodium 2,2'-biquinoline-4,4'-dicarboxylate, 2,2'-Biquinoline-4,4'-dicarboxylic acid disodium salt, Disodium 2,2'-Biquinoline-4,4'-dicarboxylate, Bicinchoninic acid disodium salt
44247.04
44247.09
44247.04EA
113
EUR
44247.04
44247.09
2,2'-Bicinchoninic acid disodium salt
2,2'-Bicinchoninic acid disodium salt
CAS No.: 979-88-4
Molecular Formula: C20H10N2O4Na2
Formula Weight: 388.29
Physical Form: Powder/chunks
MDL No.: MFCD00037500
Applications: For determination of Cu and protein assays
Formel:
C₂₀H₁₀N₂Na₂O₄ Molecular Weight: 388,29 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037500 CAS-Nummer: 979-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...