Tris(cyclopentadienyl)holmium(III) ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Unter Argon in Ampullen verpackt
44302.03
44302.06
44302.03EA
124
EUR
44302.03
44302.06
Tris(cyclopentadienyl)holmium(III) ≥98%
Tris(cyclopentadienyl)holmium(III)
Purity: 98%
CAS No.: 1272-22-6
Molecular Formula: C15H15Ho
Formula Weight: 360.21
Physical Form: Crystalline, Ampouled under argon
Sensitivity: Moisture Sensitive
Melting Point: 295°
Boiling Point: 230° subl. (vac.)
UN No.: UN3181
MDL No.: MFCD03427140
Formel:
C₁₅H₁₅Ho Molecular Weight: 360.21 g/mol Siedepunkt: 230 °C (Subl. im Vacuum) Schmelzpunkt: 295 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03427140 CAS-Nummer: 1272-22-6 EINECS: 215-047-4 UN: 3181 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...