Di-µ-bromobis(tri-tert-butylphosphine)dipalladium(I)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
44446.02
44446.04
44446.77
44446.02EA
208
EUR
44446.02
44446.04
44446.77
Di-µ-bromobis(tri-tert-butylphosphine)dipalladium(I)
Di-µ-bromobis(tri-tert-butylphosphine)dipalladium(I)
CAS No.: 185812-86-6
Molecular Formula: C24H54Br2P2Pd2
Formula Weight: 777.29
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Air Sensitive
Appearance: Dark green
Solubility: Soluble in benzene and toluene
MDL No.: MFCD04114019
Applications: Coupling reactions. Will activate aryl chloride and sterically hindered or electron rich aryl/vinyl bromides and iodides. Especially active in difficult aminations
Formel:
C₁₂H₅₄Br₂P₂Pd₂ |
MDL:
MFCD04114019 CAS-Nummer: 185812-86-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...