Scandium(III) 2,4-pentanedionate hydrate

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Scandium(III) 2,4-pentanedionate xhydrate

44448.03 44448.06
44448.03EA 113 EUR
44448.03 44448.06
Scandium(III) 2,4-pentanedionate hydrate
Scandium(III) 2,4-pentanedionate hydrate

CAS No.: 699012-88-9 Molecular Formula: C15H21O6Sc.xH2O Formula Weight: 342.26(anhy) Physical Form: Powder Odour: Odourless Melting Point: 174-186° MDL No.: MFCD00064715

Formel: C₁₅H₂₁O₆Sc·xH₂O
Schmelzpunkt: 174…186 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064715
CAS-Nummer: 699012-88-9
EINECS: 238-194-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR