Yttrium(III) trifluoroacetate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Scientific
Achtung

44597.09 44597.18
44597.09EA 72.6 EUR
44597.09 44597.18
Yttrium(III) trifluoroacetate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Yttrium(III) trifluoroacetate hydrate

Purity: 99.9% CAS No.: 304851-95-4 Molecular Formula: Y(OOCCF3)3.xH2O Formula Weight: 427.95(anhy) Physical Form: Crystalline MDL No.: MFCD05663780

Formel: C₆F₉O₆Y·xH₂O
Molecular Weight: 427,951 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD05663780
CAS-Nummer: 304851-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR