Poly(acrylic acid) 25% (w/w) in Wasser

Lieferant: Thermo Fisher Scientific

44669.22 44669.36
44669.22EA 63.3 EUR
44669.22 44669.36
Poly(acrylic acid) 25% (w/w) in Wasser
Poly(acrylic acid)

CAS No.: 9003-01-4 Molecular Formula: [-CH2CH(CO2H)-] Physical Form: Liquid Density: 1.09 MDL No.: MFCD00084394

Formel: C₃H₄O₂
Molecular Weight: 72.063 g/mol
Dichte: 1.09
Lagertemperatur: Raumtemperatur
MDL: MFCD00084394
CAS-Nummer: 9003-01-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR