Niobium(V) oxalate hydrate

Lieferant: Thermo Fisher Scientific
Achtung

44819.14 44819.22 44819.36
44819.14EA 63 EUR
44819.14 44819.22 44819.36
Niobium(V) oxalate hydrate
Niobium(V) oxalate hydrate

CAS No.: 21348-59-4 Molecular Formula: C10H5NbO20.xH2O Formula Weight: 538.04(anhy) Physical Form: Powder Odour: Odourless MDL No.: MFCD00050743

Formel: C₁₀H₅NbO₂₀·xH₂O
Molecular Weight: 538,04 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00050743
CAS-Nummer: 21348-59-4
EINECS: 244-341-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR