Iron(III) isopropoxide ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Iron(III) i-propoxide

Unter Argon in Ampullen verpackt

44873.03 44873.06 44873.14
44873.03EA 96.9 EUR
44873.03 44873.06 44873.14
Iron(III) isopropoxide ≥98%
Iron(III) isopropoxide

Purity: 98% CAS No.: 14995-22-3 Molecular Formula: C9H21FeO3 Formula Weight: 233.11 Physical Form: Solid mass, Ampouled under argon Sensitivity: Moisture Sensitive Appearance: Dark grey UN No.: UN3181 MDL No.: MFCD00070436

Formel: C₉H₂₁FeO₃
Molecular Weight: 233,109 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00070436
CAS-Nummer: 14995-22-3
UN: 3181
ADR: 4.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR