Chlorobis(cyclooctene)iridium(I) dimer (∼42,9% Ir)

Lieferant: Thermo Fisher Scientific
Achtung

44966.06 44966.03
44966.06EA 1645 EUR
44966.06 44966.03
Chlorobis(cyclooctene)iridium(I) dimer (∼42,9% Ir)
Chlorobis(cyclooctene)iridium(I) dimer

CAS No.: 12246-51-4 Molecular Formula: C32H56Cl2Ir2 Formula Weight: 896.00 MDL No.: MFCD00213465

Formel: C₃₂H₅₆Cl₂Ir₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00213465
CAS-Nummer: 12246-51-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR