Methoxy(cyclooctadiene)iridium(I) dimer (∼58% Ir)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Di-mu-methoxobis(1,5-cyclooctadiene)diiridium(I), Bis(1,5-cyclooctadiene)di-mu-methoxydiiridium(I), (1,5-Cyclooctadiene)(methoxy)iridium(I) dimer, Bis(1,5-cyclooctadiene)dimethoxydiiridium
44967.06
44967.03
44967.06EA
2161
EUR
44967.06
44967.03
Methoxy(cyclooctadiene)iridium(I) dimer (∼58% Ir)
Methoxy(cyclooctadiene)iridium(I) dimer
CAS No.: 12148-71-9
Molecular Formula: C18H30Ir2O2
Formula Weight: 662.88
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Light, Air and Moisture Sensitive
Appearance: Yellow
MDL No.: MFCD08459360
Formel:
C₁₈H₃₀Ir₂O₂ Molecular Weight: 662.87 g/mol |
MDL:
MFCD08459360 CAS-Nummer: 12148-71-9 |