Poly(styrene sulphonic acid)sodium salt, M.W. 500,000
Lieferant: Thermo Fisher Scientific
Synonyme:
Poly(sodium-p-styrenesulphonate)
45050.22
45050.14
45050.22EA
139
EUR
45050.22
45050.14
Poly(styrene sulphonic acid)sodium salt, M.W. 500,000
Poly(styrene sulphonic acid)sodium salt
CAS No.: 25704-18-1
Molecular Formula: [-CH2CH(C6H4)SO3Na)-]n
Physical Form: Powder
Appearance: White to pale brown
MDL No.: MFCD00084449
Formel:
[-CH₂CH(C₆H₄)SO₃Na)-]n Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084449 CAS-Nummer: 25704-18-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...