Tetraamminepalladium(II) hydrogen carbonate

Lieferant: Thermo Scientific
Gefahr

45082.03 45082.14 45082.06
45082.03EA 115 EUR
45082.03 45082.14 45082.06
Tetraamminepalladium(II) hydrogen carbonate
Tetraamminepalladium(II) hydrogen carbonate

CAS No.: 134620-00-1 Molecular Formula: C2H14N4O6Pd Formula Weight: 296.58 Physical Form: Crystalline Density: 2.04 Appearance: Colourless to light yellow UN No.: UN3077 MDL No.: MFCD09953449

Formel: C₂H₁₄N₄O₆Pd
Molecular Weight: 296,58 g/mol
Dichte: 2,04
Lagertemperatur: Raumtemperatur
MDL: MFCD09953449
CAS-Nummer: 134620-00-1
EINECS: 425-270-0
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR