Methylcellulose, 15 cPs
Lieferant: Thermo Scientific
Synonyme:
Cellulose methyl ether
45490.A4
45490.22
45490.36
45490.A4EA
701
EUR
45490.A4
45490.22
45490.36
Methylcellulose, 15 cPs
Methyl cellulose
CAS No.: 9004-67-5
Physical Form: Powder
Odour: Odourless
MDL No.: MFCD00081763
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00081763 CAS-Nummer: 9004-67-5 Merck Index: 14,6040 |