Silver(I) citrate hydrate
Lieferant: Thermo Scientific
|
Achtung
|
45496.06
45496.14
45496.22
45496.06EA
105
EUR
45496.06
45496.14
45496.22
Silver(I) citrate hydrate
Silver(I) citrate hydrate
CAS No.: 206986-90-5
Molecular Formula: Ag3C6H5O7.xH2O
Formula Weight: 512.70(anhy)
Physical Form: Powder
Sensitivity: Light Sensitive
Melting Point: 170° dec.
MDL No.: MFCD00150589
Formel:
C₆H₅Ag₃O₇·xH₂O Molecular Weight: 512,70 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150589 CAS-Nummer: 206986-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...