2,3,4,6-Tetra-O-benzyl-D-galactopyranose ≥98%

Lieferant: Thermo Scientific

Synonyme: (3R,4S,5S,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol, 2,3,4,6-Tetra-O-benzyl-D-galactose

45531.03 45531.06
45531.03EA 115 EUR
45531.03 45531.06
2,3,4,6-Tetra-O-benzyl-D-galactopyranose ≥98%
2,3,4,6-Tetra-O-benzyl-D-galactopyranose

Purity: 98% CAS No.: 53081-25-7 Molecular Formula: C34H36O6 Formula Weight: 540.65 Storage Temperature: -30°C to -10°C Physical Form: Crystalline Appearance: White MDL No.: MFCD00132927

Formel: C₃₄H₃₆O₆
Molecular Weight: 540,65 g/mol
MDL: MFCD00132927
CAS-Nummer: 53081-25-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR