Nickel iron molybdenum, Schuppen -200 mesh
Lieferant: Thermo Scientific
|
Gefahr
|
45610.A1
45610.30
45610.A1EA
243
EUR
45610.A1
45610.30
Nickel iron molybdenum, Schuppen -200 mesh
Nickel iron molybdenum
Molecular Formula: Ni:Fe:Mo; 81:17:2 wt%
Physical Form: Flakes
Density: 0.29
UN No.: UN3089
MDL No.: MFCD09910537
Formel:
Ni:Fe:Mo;81:17:2% Dichte: 0,29 g/mL Lagertemperatur: Raumtemperatur |
MDL:
MFCD09910537 UN: 3089 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...